missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydrochloric Acid, 6.00 N, Ricca Chemical
$91.72 - $643.03
Chemical Identifiers
| CAS | 7732-18-5 |
|---|---|
| Molecular Formula | ClH |
| Molecular Weight (g/mol) | 36.46 |
| MDL Number | MFCD00011324 MFCD00792839 |
| InChI Key | VEXZGXHMUGYJMC-UHFFFAOYSA-N |
| Synonym | hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof |
| PubChem CID | 313 |
| ChEBI | CHEBI:17883 |
| IUPAC Name | hydrogen chloride |
| SMILES | Cl |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
R3750000250
|
Ricca Chemical Company
R3750000250A |
250 mL | Poly Bottle |
Each for $134.34
|
|
||||
|
3750-16
|
Ricca Chemical Company
375016 |
500 mL | Natural Poly Bottle |
Each for $100.31
|
|
||||
|
3750-32
|
Ricca Chemical Company
375032 |
1 L | Natural Poly Bottle |
Each for $91.72
|
|
||||
|
R37500004F
|
Ricca Chemical Company
R37500004F |
4 L | Cubitainer |
Each for $223.06
|
|
||||
|
3750-1
|
Ricca Chemical Company
RI37501 |
4 L | Natural Poly Bottle |
Each for $471.81
|
|
||||
|
3750-25
|
Ricca Chemical Company
375025 |
10 L | Cubitainer |
Each for $472.06
|
|
||||
|
3750-5
|
Ricca Chemical Company
37505 |
20 L | Cubitainer |
Each for $643.03
|
|
||||
Chemical Identifiers
| 7732-18-5 | |
| 36.46 | |
| VEXZGXHMUGYJMC-UHFFFAOYSA-N | |
| 313 | |
| hydrogen chloride |
| ClH | |
| MFCD00011324 MFCD00792839 | |
| hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof | |
| CHEBI:17883 | |
| Cl |
Specifications
| 7732-18-5,7647-01-0 | |
| 64.0,37.75 | |
| 250 mL | |
| MFCD00011324 MFCD00792839 | |
| VEXZGXHMUGYJMC-UHFFFAOYSA-N | |
| hydrogen chloride | |
| 313 | |
| Laboratory | |
| Hydrochloric Acid |
| 62.0,36.27 | |
| Poly Bottle | |
| ClH | |
| hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof | |
| Cl | |
| 36.46 | |
| CHEBI:17883 | |
| 6.00N |