missing translation for 'onlineSavingsMsg'
Learn More

Hydrochloric Acid, 6.00 N, Ricca Chemical

Catalog No. 375032
Change view
Click to view available options
Quantity:
250 mL
500 mL
1 L
4 L
10 L
20 L
Packaging:
Cubitainer
Natural Poly Bottle
Poly Bottle
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
375032 1 L Natural Poly Bottle
R3750000250 250 mL Poly Bottle
375016 500 mL Natural Poly Bottle
R37500004F 4 L Cubitainer
37501 4 L Natural Poly Bottle
375025 10 L Cubitainer
37505 20 L Cubitainer
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 375032 Supplier Ricca Chemical Company Supplier No. 375032

Chemical Identifiers

CAS 7732-18-5 , 7647-01-0
Molecular Formula ClH
Molecular Weight (g/mol) 36.46
MDL Number MFCD00011324 MFCD00792839
InChI Key VEXZGXHMUGYJMC-UHFFFAOYSA-N
Synonym hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof
PubChem CID 313
ChEBI CHEBI:17883
IUPAC Name hydrogen chloride
SMILES Cl

Specifications

CAS Min % 62.0, 36.27
CAS Max % 64.0, 37.75
Packaging Natural Poly Bottle
Quantity 1 L
Grade Laboratory
Concentration or Composition (by Analyte or Components) 6.00N
Chemical Name or Material Hydrochloric Acid