missing translation for 'onlineSavingsMsg'
Learn More

Hydrochloric Acid, 6.00 N, Ricca Chemical

Supplier:  Ricca Chemical Company R37500004F

 View more versions of this product

Catalog No. R37500004F


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Hydrochloric Acid
62.0,36.27
Cubitainer
ClH
hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof
Cl
36.46
CHEBI:17883
6.00N
7732-18-5,7647-01-0
64.0,37.75
4 L
MFCD00011324 MFCD00792839
VEXZGXHMUGYJMC-UHFFFAOYSA-N
hydrogen chloride
313
Laboratory

Chemical Identifiers

7732-18-5
36.46
VEXZGXHMUGYJMC-UHFFFAOYSA-N
313
hydrogen chloride
ClH
MFCD00011324 MFCD00792839
hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof
CHEBI:17883
Cl
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions