missing translation for 'onlineSavingsMsg'
Learn More

Hydrochloric Acid, 6.00 N, Ricca Chemical

Packaged in Glass

$134.87 - $366.33

Chemical Identifiers

CAS 7732-18-5
Molecular Formula ClH
Molecular Weight (g/mol) 36.46
MDL Number MFCD00011324 MFCD00792839
InChI Key VEXZGXHMUGYJMC-UHFFFAOYSA-N
Synonym hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof
PubChem CID 313
ChEBI CHEBI:17883
IUPAC Name hydrogen chloride
SMILES Cl
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
37501-32
SDS Accessories View Documents
Ricca Chemical Company
3750132
1 L Amber Glass Bottle
Each for $134.87
Only null left
 
3750-15PT
SDS Accessories View Documents
Ricca Chemical Company
375015PT
2.5 L Safety-coated Glass Bottle
Each for $256.60
Only null left
 
3750-11
SDS View Documents
Ricca Chemical Company
375011
4 L Amber Glass Bottle
Each for $366.33
Only null left
 
Description

Description

Specifications

Chemical Identifiers

7732-18-5
36.46
VEXZGXHMUGYJMC-UHFFFAOYSA-N
313
hydrogen chloride
ClH
MFCD00011324 MFCD00792839
hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof
CHEBI:17883
Cl

Specifications

7732-18-5,7647-01-0
64.0,37.75
1 L
MFCD00011324 MFCD00792839
VEXZGXHMUGYJMC-UHFFFAOYSA-N
hydrogen chloride
313
36.46
6.00N
62.0,36.27
Amber Glass Bottle
ClH
hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof
Cl
36.46
CHEBI:17883
Laboratory
Hydrochloric Acid
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'