missing translation for 'onlineSavingsMsg'
Learn More

Hydrochloric Acid, 6.00 N, Ricca Chemical

Catalog No. 3750132
Change view
Click to view available options
Quantity:
1 L
2.5 L
4 L
Packaging:
Amber Glass Bottle
Safety-coated Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
3750132 1 L Amber Glass Bottle
375015PT 2.5 L Safety-coated Glass Bottle
375011 4 L Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 3750132 Supplier Ricca Chemical Company Supplier No. 3750.132
Edge

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Packaged in Glass

Chemical Identifiers

CAS 7732-18-5 , 7647-01-0
Molecular Formula ClH
Molecular Weight (g/mol) 36.46
MDL Number MFCD00011324 MFCD00792839
InChI Key VEXZGXHMUGYJMC-UHFFFAOYSA-N
Synonym hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof
PubChem CID 313
ChEBI CHEBI:17883
IUPAC Name hydrogen chloride
SMILES Cl

Specifications

CAS Min % 62.0, 36.27
CAS Max % 64.0, 37.75
Packaging Amber Glass Bottle
Quantity 1 L
Formula Weight 36.46
Grade Laboratory
Concentration or Composition (by Analyte or Components) 6.00N
Chemical Name or Material Hydrochloric Acid