missing translation for 'onlineSavingsMsg'
Learn More

Hydrochloric Acid, 6.00 N, Ricca Chemical

Packaged in Glass

Supplier:  Ricca Chemical Company 375015PT

 View more versions of this product

Catalog No. 3750-15PT


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Hydrochloric Acid
62.0,36.27
Safety-coated Glass Bottle
ClH
hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof
Cl
36.46
CHEBI:17883
Laboratory
7732-18-5,7647-01-0
64.0,37.75
2.5 L
MFCD00011324 MFCD00792839
VEXZGXHMUGYJMC-UHFFFAOYSA-N
hydrogen chloride
313
36.46
6.00N

Chemical Identifiers

7732-18-5
36.46
VEXZGXHMUGYJMC-UHFFFAOYSA-N
313
hydrogen chloride
ClH
MFCD00011324 MFCD00792839
hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof
CHEBI:17883
Cl
Product Suggestions

Product Suggestions

Videos
SDS Documents

Documents

Accessories
Promotions

Promotions