missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydrochloric acid, 5.0N Standardized Solution
Description
Hydrochloric acid, 5.0N standardized solution is used for volumetric analysis in laboratories.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Vapor Pressure | 23 hPa (17mm Hg) at 20°C |
| Physical Form | Liquid |
| Color | Colorless to Yellow |
| Solubility | Miscible with water, methanol,ethanol and ether |
| IUPAC Name | hydrogen chloride |
| Quantity | 20 L |
| Formula Weight | 36.46 |
| Concentration or Composition (by Analyte or Components) | Hydrochloric acid: 41.3%; Water: 58.7% |
| Chemical Name or Material | Hydrochloric acid |
Safety and Handling
| Hazard Category | H290-H314-H335 |
| Precautionary Statement | P234-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P390-P501c |
| DOTInformation | Hazard Class: 8; Packaging Group: II |
| EINECSNumber | 231-595-7 |
| RTECSNumber | MW4025000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only