missing translation for 'onlineSavingsMsg'
Learn More

Hydrochloric Acid, 37%, NF, 36.5-38%, Spectrum™ Chemical

HY104, 7647-01-0, HCl

$407.63 - $7793.20

Chemical Identifiers

CAS 7647-01-0
Molecular Formula ClH
Molecular Weight (g/mol) 36.46
MDL Number MFCD00011324 MFCD00792839
InChI Key VEXZGXHMUGYJMC-UHFFFAOYSA-N
IUPAC Name hydrogen chloride
SMILES Cl
View More Specs

Products 10
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
18603190
View Documents
Spectrum Chemical Mfg Cor
HY104500MLGL
500 mL Amber Glass Bottle
Each for $407.63
Only null left
 
18603196
View Documents
Spectrum Chemical Mfg Cor
HY10425LTGL
2.5 L Glass Bottle
Each for $596.80
Only null left
 
18603187
View Documents
Spectrum Chemical Mfg Cor
HY10425LTPV
2.5 L Poly-Coated Glass Bottle
Each for $655.27
Only null left
 
18603191
View Documents
Spectrum Chemical Mfg Cor
HY104500MLGLCS6
6 x 500 mL Amber Glass Bottle
Case of 6 Each for $1,614.17
Only null left
 
18603192
View Documents
Spectrum Chemical Mfg Cor
HY1044LTPV
4 L Poly-Coated Glass Bottle
Each for $756.80
Only null left
 
18603195
View Documents
Spectrum Chemical Mfg Cor
HY10425LTGLCS4
4 x 2.5 L Glass Bottle
Case of 4 Each for $1,454.03
Only null left
 
18603188
View Documents
Spectrum Chemical Mfg Cor
HY10425LTPVCS4
4 x 2.5 L Poly-Coated Glass Bottle
Case of 4 Each for $1,708.63
Only null left
 
18603193
View Documents
Spectrum Chemical Mfg Cor
HY1044LTPVCS4
4 x 4 L Poly-Coated Glass Bottle
Case of 4 Each for $2,228.47
Only null left
 
18603189
View Documents
Spectrum Chemical Mfg Cor
HY10420LT
20 L Poly Drum
Each for $1,598.40
Only null left
 
18603194
View Documents
Spectrum Chemical Mfg Cor
HY104200LTBL
200 L Drum
Each for $7,793.20
Only null left
 
Description

Description

Spectrum™ Chemical Hydrochloric Acid, 37 Percent, NF is an excellent acidifying reagent. All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Specifications

Chemical Identifiers

7647-01-0
36.46
VEXZGXHMUGYJMC-UHFFFAOYSA-N
Cl
ClH
MFCD00011324 MFCD00792839
hydrogen chloride

Specifications

7647-01-0,7732-18-5
Amber Glass Bottle
ClH
UN1789
Cl
36.46
NF
36 to 38%,62 to 64%
500 mL
MFCD00011324 MFCD00792839
VEXZGXHMUGYJMC-UHFFFAOYSA-N
hydrogen chloride
36.5 to 38%
Hydrochloric Acid
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'