missing translation for 'onlineSavingsMsg'
Learn More

Hydrochloric Acid, 37%, NF, 36.5-38%, Spectrum™ Chemical

HY104, 7647-01-0, HCl

Supplier:  Spectrum Chemical Mfg Cor HY10425LTGL

 View more versions of this product

Catalog No. 18603196


Only null left
Explore available promotions

Description

Description

Spectrum™ Chemical Hydrochloric Acid, 37 Percent, NF is an excellent acidifying reagent. All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Specifications

Specifications

Hydrochloric Acid
36 to 38%,62 to 64%
2.5 L
MFCD00011324 MFCD00792839
VEXZGXHMUGYJMC-UHFFFAOYSA-N
hydrogen chloride
36.5 to 38%
7647-01-0,7732-18-5
Glass Bottle
ClH
UN1789
Cl
36.46
NF

Chemical Identifiers

7647-01-0
36.46
VEXZGXHMUGYJMC-UHFFFAOYSA-N
Cl
ClH
MFCD00011324 MFCD00792839
hydrogen chloride
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions