missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydrochloric Acid, 37%, NF, 36.5-38%, Spectrum™ Chemical
HY104, 7647-01-0, HCl
Supplier: Spectrum Chemical Mfg Cor HY104500MLGL
Description
Spectrum™ Chemical Hydrochloric Acid, 37 Percent, NF is an excellent acidifying reagent. All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Specifications
| Hydrochloric Acid | |
| 36 to 38%,62 to 64% | |
| 500 mL | |
| MFCD00011324 MFCD00792839 | |
| VEXZGXHMUGYJMC-UHFFFAOYSA-N | |
| hydrogen chloride | |
| 36.5 to 38% |
| 7647-01-0,7732-18-5 | |
| Amber Glass Bottle | |
| ClH | |
| UN1789 | |
| Cl | |
| 36.46 | |
| NF |
Chemical Identifiers
| 7647-01-0 | |
| 36.46 | |
| VEXZGXHMUGYJMC-UHFFFAOYSA-N | |
| Cl |
| ClH | |
| MFCD00011324 MFCD00792839 | |
| hydrogen chloride |