missing translation for 'onlineSavingsMsg'
Learn More

Hydrochloric acid, 1.0N Standardized Solution

Catalog No. AA35640M4
Change view
Click to view available options
Quantity:
1 L
4 L
20 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA35640M4 20 L
AA35640K2 1 L
AA35640K7 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA35640M4 Supplier Thermo Scientific Chemicals Supplier No. 035640M4

CAS: 7647-01-0 | ClH | 36.46 g/mol

Hydrochloric acid, 1.0N standardized solution is used for volumetric analysis in laboratories.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7647-01-0 , 7732-18-5
Molecular Formula ClH
Molecular Weight (g/mol) 36.46
MDL Number MFCD00011324 MFCD00792839
InChI Key VEXZGXHMUGYJMC-UHFFFAOYSA-N
PubChem CID 313
ChEBI CHEBI:17883
SMILES Cl

Specifications

Vapor Pressure 23 hPa (17mm Hg) at 20°C
Physical Form Liquid
Color Colorless to Yellow
Solubility Miscible with water, methanol,ethanol and ether
IUPAC Name chlorane
Quantity 20 L
Formula Weight 36.46
Concentration or Composition (by Analyte or Components) Hydrochloric acid: 8.26%; Water: 91.74%
Chemical Name or Material Hydrochloric acid
Hazard Category H290-H315-H318
Precautionary Statement P234-P264b-P280-P302+P352-P305+P351+P338-P310-P332+P313-P362-P390
DOTInformation Hazard Class: 8; Packaging Group: III
EINECSNumber 231-595-7
RTECSNumber MW4025000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only