missing translation for 'onlineSavingsMsg'
Learn More

Hydrochloric Acid, 0.773 N, Ricca Chemical

Chemical Identifiers

CAS 7647-01-0 , 7732-18-5
Molecular Formula ClH
Molecular Weight (g/mol) 36.46
MDL Number MFCD00011324
InChI Key VEXZGXHMUGYJMC-UHFFFAOYSA-N
Synonym hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof
PubChem CID 313
ChEBI CHEBI:17883
IUPAC Name hydrogen chloride
SMILES [H+].[Cl-]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
367332
View Documents Promotion Details
N/A 1 L Natural Poly Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
367325
View Documents Promotion Details
N/A 10 L Cubitainer N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

7647-01-0 , 7732-18-5
36.46
VEXZGXHMUGYJMC-UHFFFAOYSA-N
313
hydrogen chloride
ClH
MFCD00011324
hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof
CHEBI:17883
[H+].[Cl-]

Specifications

7647-01-0 , 7732-18-5
95.0, 7.2
MFCD00011324
VEXZGXHMUGYJMC-UHFFFAOYSA-N
hydrogen chloride
313
Hydrochloric Acid
91.0, 6.92
ClH
hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof
[H+].[Cl-]
36.46
CHEBI:17883
Videos
SDS
Documents

Documents