missing translation for 'onlineSavingsMsg'
Learn More

Hydrochloric Acid, 0.773 N, Ricca Chemical

Supplier:  Ricca Chemical Company 36732.5

Quantity 10 L
Packaging Cubitainer

 View more versions of this product

Catalog No. 367325

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Chemical Identifiers

7647-01-0 , 7732-18-5
36.46
VEXZGXHMUGYJMC-UHFFFAOYSA-N
313
hydrogen chloride
ClH
MFCD00011324
hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof
CHEBI:17883
[H+].[Cl-]

Specifications

Hydrochloric Acid
91.0, 6.92
Cubitainer
ClH
hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof
[H+].[Cl-]
36.46
CHEBI:17883
7647-01-0 , 7732-18-5
95.0, 7.2
10 L
MFCD00011324
VEXZGXHMUGYJMC-UHFFFAOYSA-N
hydrogen chloride
313
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions