missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydrochloric Acid, 0.773 Normal, Reagents
Spécifications
Spécifications
| pH | <1 |
| Color | Colorless |
| Boiling Point | 100°C |
| Packaging | Cubitainer |
| Specific Gravity | 1.0137 |
| Quantity | 10 L |
| Physical Form | Liquid |
| Grade | Reagent |
| Chemical Name or Material | Hydrochloric Acid, 0.773 Normal |