missing translation for 'onlineSavingsMsg'
Learn More

Hydrochloric Acid, 0.200 N (N/5) in Isopropyl Alcohol, Ricca Chemical

Catalog No. 380232
Change view
Click to view available options
Quantity:
1 L
4 L
Packaging:
Natural Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
380232 1 L Natural Poly Bottle
38021 4 L Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 380232 Supplier Ricca Chemical Company Supplier No. 380232
Only null left

Chemical Identifiers

CAS 67-63-0 , 7732-18-5 , 7647-01-0

Specifications

Color Colorless
Concentration 0.200Normal
Packaging Natural Poly Bottle
Solvent or Matrix Isopropyl Alcohol
Quantity 1 L
Molecular Formula ClH
Physical Form Liquid
MDL Number MFCD00011324 MFCD00792839
Synonym hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof
InChI Key VEXZGXHMUGYJMC-UHFFFAOYSA-N
SMILES Cl
IUPAC Name hydrogen chloride
Molecular Weight (g/mol) 36.46
PubChem CID 313
ChEBI CHEBI:17883
Grade Laboratory
Chemical Name or Material Hydrochloric Acid
Show More Show Less