missing translation for 'onlineSavingsMsg'
Learn More

Hydrochloric Acid, 0.200 N (N/5) in Isopropyl Alcohol, Ricca Chemical

Catalog No. 38021
Change view
Click to view available options
Quantity:
1 L
4 L
Packaging:
Natural Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
38021 4 L Natural Poly Bottle
380232 1 L Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 38021 Supplier Ricca Chemical Company Supplier No. 38021
Only null left

Chemical Identifiers

CAS 67-63-0 , 7732-18-5 , 7647-01-0

Specifications

Packaging Natural Poly Bottle
Quantity 4 L
Molecular Formula Cl2Pd
MDL Number MFCD00003558
Synonym palladium chloride, palladium ii chloride, palladium dichloride, palladous chloride, pdcl2, palladium 2+ chloride, palladium chloride pdcl2, enplate activator 440
InChI Key PIBWKRNGBLPSSY-UHFFFAOYSA-L
SMILES [Cl-].[Cl-].[Pd++]
IUPAC Name palladium(2+) dichloride
Molecular Weight (g/mol) 177.32
PubChem CID 24290
ChEBI CHEBI:53434
Grade Laboratory
Chemical Name or Material Hydrochloric Acid, 0.200 (±0.0005) Normal (N/5)
Show More Show Less