missing translation for 'onlineSavingsMsg'
Learn More

Hydrobromic Acid (OPTIMA™), Fisher Chemical™

Catalog No. p-326250
Change view
Click to view available options
Quantity:
250 mL
500 mL
1 L
2 L
Packaging:
FEP bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
A471250 250 mL FEP bottle
A471500 500 mL FEP bottle
A4711 1 L FEP bottle
A4712 2 L FEP bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. A471250 Supplier Seastar Chemicals Inc Supplier No. S020801TFIF01
Only null left

Chemical Identifiers

CAS 10035-10-6 , 7732-18-5
Molecular Formula BrH
Molecular Weight (g/mol) 80.91
MDL Number MFCD00011323
InChI Key CPELXLSAUQHCOX-UHFFFAOYSA-N
Synonym hydrogen bromide, hydrobromic acid, bromwasserstoff, broomwaterstof, bromowodor, acido bromidrico, acide bromhydrique, anhydrous hydrobromic acid, bromowodor polish, hydrobromide
PubChem CID 260
ChEBI CHEBI:47266
IUPAC Name hydrogen bromide
SMILES Br

Specifications

pH <1
Color Yellow
Melting Point -11°C
Boiling Point 126°C
Density 1.480g/cm³
Packaging FEP bottle
Vapor Pressure 10 mm Hg at 25°C
Quantity 250 mL
Physical Form Liquid
Merck Index 15, 4816
Formula Weight 80.91
Grade OPTIMA
Chemical Name or Material Hydrobromic Acid, 44–49%
Show More Show Less