missing translation for 'onlineSavingsMsg'
Learn More

Hydrazobenzene 98.0+%, TCI America™

Catalog No. H018025G
Change view
Click to view available options
Quantity:
25 g
100 g
Catalog No. Quantity
H018025G 25 g
H0180100G 100 g
2 options

Catalog No. H018025G

Supplier: TCI America H018025G

Only null left

Chemical Identifiers

CAS 122-66-7
Molecular Formula C12H12N2
Molecular Weight (g/mol) 184.242
MDL Number MFCD00003012
InChI Key YBQZXXMEJHZYMB-UHFFFAOYSA-N
Synonym hydrazobenzene, 1,2-diphenyldiazane, hydrazine, 1,2-diphenyl, n,n'-bianiline, n,n'-diphenylhydrazine, hydrazobenzen, benzene, hydrazodi, sym-diphenylhydrazine, rcra waste number u109, hydrazine, diphenyl
PubChem CID 31222
IUPAC Name 1,2-diphenylhydrazine
SMILES C1=CC=C(C=C1)NNC2=CC=CC=C2

Specifications

Melting Point 127°C
Color White-Yellow
Quantity 25 g
UN Number 3077
Formula Weight 184.24
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Hydrazobenzene
EINECSNumber (3)-2755
RTECSNumber MW2625000
TSCA Yes
Recommended Storage Refrigerator