missing translation for 'onlineSavingsMsg'
Learn More

Hydrazine Sulfate, CP, Spectrum™ Chemical

Catalog No. 18610833
Change view
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
25 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18610833 500 g Amber Glass Bottle
18610831 2.5 kg Amber Glass Bottle
18610830 12 kg Poly Pail
18610832 25 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18610833 Supplier Spectrum Chemical Mfg Cor Supplier No. H1019500GM
Get Up to $500 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 10034-93-2

Specifications

CAS Max % 1
Packaging Amber Glass Bottle
Quantity 500 g
Grade Chemically Pure

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.