missing translation for 'onlineSavingsMsg'
Learn More

Hydrazine Standard, Certified, 1000ppm ±10ppm (1mL = 1mg N2H4), LabChem™

Catalog No. LC149301
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
LC149301 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. LC149301 Supplier LabChem Supplier No. LC149301
Edge

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 5341-61-7 , 7647-01-0 , 7732-18-5
Molecular Formula ClH5N2
Molecular Weight (g/mol) 68.50
MDL Number MFCD00064543
InChI Key BIVUUOPIAYRCAP-UHFFFAOYSA-N
PubChem CID 17548
IUPAC Name hydrogen hydrazine chloride
SMILES [H+].[Cl-].NN

Specifications

Packaging Poly Bottle
CAS Max % 99.17, 0.5, 0.33
Identification Passes Test
Density 1g/mL
Physical Form Liquid
Concentration 1000 ppm ±10 ppm (1mL = 1mg N2H4)
Linear Formula N2H4·2HCl
UN Number UN1789
Color Colorless
Solubility Soluble in water
Quantity 500 mL
Formula Weight 104.97
Grade Certified
Chemical Name or Material Hydrazine Standard, 1000ppm (1mL = 1mg N2H4)
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement
May cause an allergic skin reaction.
May cause cancer.
Harmful to the environment with long lasting effects.
Precautionary Statement GHS P Statement
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
Do not breathe mist, vapors, spray.
Wear protective gloves, eye protection.
Contaminated work clothing should not be allowed out of the workplace.
If exposed or concerned: Get medical advice/attention.
If on skin (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
If skin irritation or rash occurs: Get medical advice/attention.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.
Recommended Storage Room Temperature