missing translation for 'onlineSavingsMsg'
Learn More

Hydrazine Standard, 1000 ppm N2H4, Ricca Chemical

Catalog No. 355316
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
355316 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 355316 Supplier Ricca Chemical Company Supplier No. 355316

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 5341-61-7 , 7647-01-0 , 7732-18-5
Molecular Formula ClH5N2
Molecular Weight (g/mol) 68.50
MDL Number MFCD00064543
InChI Key BIVUUOPIAYRCAP-UHFFFAOYSA-N
PubChem CID 17548
IUPAC Name hydrogen hydrazine chloride
SMILES [H+].[Cl-].NN

Specifications

Packaging Amber Glass Bottle
CAS Min % 97.0, 1.14, 0.32
CAS Max % 100.0, 1.19, 0.33
Equivalence 1 mL = 1 mg N
Physical Form Liquid
Color Colorless
pH <1
Melting Point 0.0°C
Boiling Point 100°C
For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
Solubility Miscible with water
Solution Type Calibration Standard
Type Analytical Standard
Quantity 500 mL
Grade Laboratory
Chemical Name or Material Hydrazine Standard
Show More Show Less