missing translation for 'onlineSavingsMsg'
Learn More

Hydrazine Standard, 100 ppm N2H4, Ricca Chemical

Catalog No. 35524
Change view
Click to view available options
Quantity:
120 mL
500 mL
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
35524 120 mL Amber Glass Bottle
355216 500 mL Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 35524 Supplier Ricca Chemical Company Supplier No. 35524

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 5341-61-7 , 7647-01-0 , 7732-18-5
Molecular Formula ClH5N2
Molecular Weight (g/mol) 68.50
MDL Number MFCD00064543
InChI Key BIVUUOPIAYRCAP-UHFFFAOYSA-N
PubChem CID 17548
IUPAC Name hydrogen hydrazine chloride
SMILES [H+].[Cl-].NN

Specifications

Packaging Amber Glass Bottle
CAS Min % 97.0, 1.14, 0.03
CAS Max % 100.0, 1.19, 0.03
Equivalence 1 mL = 0.1 mg N
Physical Form Liquid
Color Colorless
Boiling Point 100°C
Quantity 120 mL
Chemical Name or Material Hydrazine Standard