missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HYDRANAL™-Water Standard KF-Oven, 150-160°C, Solstice
Description
Advantages of HYDRANAL-Water Standards:
- Broad product range for volumetric and coulometric Karl Fischer applications, as well as the indirect oven application
- Manufactured according to ISO 9001 requirements
- Report of Analysis showing the exact water content and expanded uncertainty
Verified against external standards such as NIST SRM 2890 or NMIJ CRM 4222
- Long shelf life (up to five years)
- Convenient packaging
- Supplied with detailed instruction for use
Specifications
Specifications
| Chemical Name or Material | HYDRANAL™-Water Standard KF-Oven, 150-160°C |
| CAS | 64044-51-5 |
| Quantity | 10 g |
| UN Number | NONH for all modes of transport |