missing translation for 'onlineSavingsMsg'
Learn More

HYDRANAL™ - Water Standard 10.0, Standard for Karl Fischer titration (water content 10 mg/g = 1.0%), Solstice

Catalog No. 60050009
Change view
Click to view available options
Quantity:
6 x (10 x 8 mL)
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60050009 6 x (10 x 8 mL)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 60050009 Supplier Solstice Supplier No. 348496X80ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 108-32-7 | C8H10 | 106.17 g/mol

Chemical Identifiers

Molecular Formula C8H10
Molecular Weight (g/mol) 106.17
MDL Number MFCD00008519
InChI Key CTQNGGLPUBDAKN-UHFFFAOYSA-N
Synonym 1,2-Propanediol cyclic carbonate, 4-Methyl-1,3-dioxolan-2-one
PubChem CID 962
ChEBI CHEBI:15377
IUPAC Name 1,2-xylene
SMILES CC1=CC=CC=C1C

Specifications

CAS 108-32-7, 71-36-3, 95-47-6, 108-32-7
Boiling Point 109°C
CAS Min % ≥20.0000%
CAS Max % <30.0000%
Density 1.00 g/mL at 20 °C
Quantity 6 x (10 x 8 mL)
Flash Point 32°C
Packaging Ampoule
UN Number UN1993
Grade Karl Fischer
Assay effective water
Chemical Name or Material HYDRANAL™ - Water Standard 10.0, Standard for Karl Fischer titration (water content 10 mg/g = 1.0%), exact value on report of analysis
Show More Show Less
ShelfLife 1800 days from date of manufacture
DOTInformation Transport Hazard Class: 3; Packing Group:3: III; Proper Shipping Name: Flammable liquids, n.o.s.
Recommended Storage Room Temp