missing translation for 'onlineSavingsMsg'
Learn More

HYDRANAL™ - Titrant 2, Reagent for volumetric two-component Karl Fischer titration, Solstice

Catalog No. 60046457
Change view
Click to view available options
Quantity:
500 mL
1 L
6 x 500 mL
6 x 1 L
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60046457 6 x 500 mL
6003225 500 mL
6003223 1 L
60046456 6 x 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 60046457 Supplier Honeywell Inc Supplier No. 348116X500MLUS

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 7553-56-2

Specifications

Chemical Name or Material HYDRANAL™ - Titrant 2
Name Note Reagent for volumetric two-component Karl Fischer titration
Boiling Point 65°C
CAS Min % ≥90.0000%, ≥1.0000%
Density ∼0.80 g/mL
CAS 7553-56-2
Quantity 6 x 500 mL
CAS Max % <5.0000%
Flash Point 11°C
Packaging Glass Bottle
UN Number UN1230
Synonym Methyl alcohol
Grade Karl Fischer
Show More Show Less