missing translation for 'onlineSavingsMsg'
Learn More

HYDRANAL™ - Titrant 2, Reagent for volumetric two-component Karl Fischer titration, Honeywell™ Fluka™

Catalog No. 6003225
Change view
Click to view available options
Quantity:
500 mL
1 L
6 x 500 mL
6 x 1 L
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
6003225 500 mL
6003223 1 L
60046457 6 x 500 mL
60046456 6 x 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 6003225 Supplier Honeywell Inc Supplier No. 34811500MLUS
Only null left

Specifications

Chemical Name or Material HYDRANAL™-Titrant 2
Density 0.80g/mL
Quantity 500 mL
Flash Point 11°C (51.8°F)
UN Number UN1230