missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HYDRANAL™-Solvent CM, Medium for volumetric two-component Karl Fischer titration in non-polar substances, fats and oils (chloroform-methanol-based), Solstice
Description
Advantages of Hydranal two-component reagents:
- High titration speed
- Ideal accuracy for small amounts of water
- High buffer capacity
- Exact and stable titer
- E-type reagents: reduced toxicity compared to methanol
- Long shelf life (three years for titrants, five years for solvents)
Improved Composition
In two-component reagents the Karl Fisher reactants are separated into two solutions: the titrant and the solvent. Hydranal-Titrant contains iodine dissolved in an alcohol with a precisely defined concentration. Hydranal-Solvent is an alcoholic solution of sulfur dioxide and imidazole. The alcohol is either methanol for standard reagents or ethanol for E-type reagents.
Specifications
Specifications
| Boiling Point | 60 to 65°C ( 1.013 hPa) |
| CAS Min % | ≥70.00, ≥20.00 |
| CAS Max % | <90.00, <5.00 |
| Density | 1.27 g/cm3 (20°C) |
| Physical Form | Liquid |
| Quantity | 1 L |
| Flash Point | 100°C |
| Packaging | Polycoated Bottle |
| Grade | Two Component Volumetry |
Safety and Handling
| Hazard Category | Danger |
| Hazard Statement | H372+H370+H360+H351+H302+H332+H315+H319+H336 |
| Precautionary Statement | P280+P271+P270+P264+P260+P201+P202 |
| Recommended Storage | Room Temperature |