missing translation for 'onlineSavingsMsg'
Learn More

HYDRANAL™ - CRM Sodium tartrate dihydrate, Certified reference material for Karl Fischer titration (water content ∼15.66%), Honeywell Fluka™

Catalog No. 60050000
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60050000 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 60050000 Supplier Honeywell Chemicals Supplier No. 3442410G
Only null left

Certified Reference Material for Karl Fischer titration (water content ∼15.66%, exact value on certificate of analysis)

Advantages of HYDRANAL- CRM Water Standards:

  • For volumetric and coulometric Karl Fischer applications
  • Manufactured according to ISO 17034 requirements
  • Certificate of Analysis showing the exact water content, expanded uncertainty plus comprehensive product information and instruction for use
  • Lot-related stability and homogeneity studies
  • Verified against external standards such as NIST SRM 2890 or NMIJ CRM 4222
  • Convenient packaging

Chemical Identifiers

Molecular Formula C4H8Na2O8
Molecular Weight (g/mol) 230.08
MDL Number MFCD00150035
InChI Key FGJLAJMGHXGFDE-UDMGOBQRNA-L
PubChem CID 131855972
IUPAC Name (2R,3R)-2,3-dihydroxybutanedioic acid;sodium;dihydrate
SMILES O.O.[Na+].[Na+].O[C@H]([C@@H](O)C([O-])=O)C([O-])=O

Specifications

Density 1.820 g/cm3
CAS 6106-24-7
Quantity 10 g
CAS Max % ≤100.0000%
Packaging Glass Bottle
Grade Certified Reference Material
Assay effective water volumetric
Chemical Name or Material HYDRANAL™ - CRM Sodium tartrate dihydrate, Certified reference material for Karl Fischer titration (water content ∼15.66%)
ShelfLife 1080 days from date of manufacture