missing translation for 'onlineSavingsMsg'
Learn More

HYDRANAL™ - Coulomat A, Reagent for coulometric KF titration (anolyte solution), preferred for cells with diaphragm, Honeywell Fluka™

Catalog No. 60046449
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60046449 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 60046449 Supplier Honeywell Inc Supplier No. 348076X500MLUS
Only null left

Specifications

Chemical Name or Material HYDRANAL™ - Coulomat A
Name Note Reagent for coulometric KF titration (anolyte solution), preferred for cells with diaphragm
Boiling Point 59°C
CAS Min % ≥5.0000%, ≥1.0000%, ≥30.0000%
CAS Max % <20.0000%, <5.0000%, <10.0000%
CAS 288-32-4, 68007-08-9, 67-66-3, 7446-09-5
Density 1.070 g/cm3
Quantity 500 mL
Flash Point 11°C
Packaging Glass Bottle
UN Number UN1992
Synonym Methyl alcohol
Grade Coulometric KF Titration
Show More Show Less