missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva HyClone™ SFM4CHO™ Media
Designed for high performance in a variety of culture vessels, including bioreactors. HyClone™ SFM4CHO™ Media is a protein-free formulation that contains no components of bovine origin.
Supplier: Cytiva SH3054802
Description
- Formulated using proprietary lipid complexing process for enhanced stability and growth promotion of various CHO cell lines
Specifications
| Liquid | |
| 0.2 μm Sterile-filtered | |
| No Glutamine, No Phenol Red, No HEPES | |
| cGMP | |
| SFM4CHO media |
| SFM4CHO Liquid Media | |
| Sodium Bicarbonate, Poloxamer 188 | |
| 1000 mL | |
| 2° to 8°C, away from light |