Learn More
Cytiva HyClone™ SFM4CHO™ Media
Designed for high performance in a variety of culture vessels, including bioreactors. HyClone™ SFM4CHO™ Media is a protein-free formulation that contains no components of bovine origin.
$128.34 - $382.06
| Catalog Number | Mfr. No. | Quantity | With Additives | Without Additives | Form | Price | Quantity & Availability | ||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | With Additives | Without Additives | Form | Price | Quantity & Availability | ||
|
SH3054902
|
Cytiva
SH30549.02 |
1000 mL | Glutamine, Sodium Bicarbonate, Poloxamer 188 | No Phenol Red, No HEPES | Liquid |
Each for $128.34
|
|
||
|
SH3051801
|
Cytiva
SH30518.01 |
5 L | No Glutamine, No Sodium Bicarbonate, No Poloxamer 188, No Phenol Red, No HEPES | Powder |
Each for $382.06
|
|
|||
|
SH3054802
|
Cytiva
SH30548.02 |
1000 mL | Sodium Bicarbonate, Poloxamer 188 | No Glutamine, No Phenol Red, No HEPES | Liquid |
Each for $129.56
|
|
||
Description
- Formulated using proprietary lipid complexing process for enhanced stability and growth promotion of various CHO cell lines
Specifications
| cGMP | |
| SFM4CHO media |
Safety and Handling
ShelfLife : 12 Months