missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HyClone™ SFM4CHO™ Media

Catalog No. SH3054902
Change view
Click to view available options
Quantity:
1000 mL
5 L
With Additives:
Glutamine, Sodium Bicarbonate, Poloxamer 188
Sodium Bicarbonate, Poloxamer 188
Without Additives:
No Glutamine, No Phenol Red, No HEPES
No Glutamine, No Sodium Bicarbonate, No Poloxamer 188, No Phenol Red, No HEPES
No Phenol Red, No HEPES
Form:
Liquid
Powder
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity With Additives Without Additives Form
SH3054902 1000 mL Glutamine, Sodium Bicarbonate, Poloxamer 188 No Phenol Red, No HEPES Liquid
SH3051801 5 L - No Glutamine, No Sodium Bicarbonate, No Poloxamer 188, No Phenol Red, No HEPES Powder
SH3054802 1000 mL Sodium Bicarbonate, Poloxamer 188 No Glutamine, No Phenol Red, No HEPES Liquid
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. SH3054902 Supplier Cytiva Supplier No. SH30549.02
NLSU: Get 1 Consumable Product When You Buy 1 Obtenez 1 produit consommable à l’achat d’un produit
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Designed for high performance in a variety of culture vessels, including bioreactors. HyClone™ SFM4CHO™ Media is a protein-free formulation that contains no components of bovine origin.

  • Formulated using proprietary lipid complexing process for enhanced stability and growth promotion of various CHO cell lines

Specifications

Form Liquid
Product Type SFM4CHO Liquid Media
Sterilization Method 0.2 μm Sterile-filtered
With Additives Glutamine, Sodium Bicarbonate, Poloxamer 188
Without Additives No Phenol Red, No HEPES
Quantity 1000 mL
Certifications/Compliance cGMP
Content And Storage 2° to 8°C, away from light
Description SFM4CHO media