missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva HyClone™ SFM4CHO™ Media
Designed for high performance in a variety of culture vessels, including bioreactors. HyClone™ SFM4CHO™ Media is a protein-free formulation that contains no components of bovine origin.
Supplier: Cytiva SH30518.01
Description
- Formulated using proprietary lipid complexing process for enhanced stability and growth promotion of various CHO cell lines
Chemical Identifiers
| cGMP | |
| SFM4CHO media |
Specifications
| Powder | |
| No Glutamine, No Sodium Bicarbonate, No Poloxamer 188, No Phenol Red, No HEPES | |
| cGMP | |
| SFM4CHO media |
| Non-sterile | |
| 5 L | |
| 2° to 8°C, away from light and moisture |
Safety and Handling
ShelfLife : 24 Months