missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HyClone™ Irradiated Calf Serum, U.S. Origin

Catalog No. SH3007302IR
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SH3007302IR 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SH3007302IR Supplier Cytiva Supplier No. SH30073.02IR

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Collected by venipuncture from formula-fed veal calves, which produce exceptionally high levels of transferrin

  • Age of animal at time of collection ranges from 16 to 24 weeks
  • Low in antibodies and high in growth factors
  • Virus panel testing according to 9 CFR 113.53
  • Sourced in the USA with complete traceability back to original source
  • Cost-effective alternative to FBS

Specifications

Age 16 to 24 weeks
Country of Origin United States
Product Type Calf Bovine Serum
Quantity 100 mL
Sterilization Method Sequential-filtered, 100 nm
Processing Irradiated
Tested For Virus Panel Testing According to 9 CFR 113.53
Heat Inactivated No
Product Line HyClone