missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva HyClone™ FetalClone™ III Serum (U.S.)
Designed for fibroblast applications, supplement cell culture growth with U.S.-sourced HyClone™ FetalClone™ III Serum, the most widely applicable fetal bovine serum (FBS) alternative.
Supplier: Cytiva SH30109.03
Description
- Economical alternative to FBS
- Performance comparable with FBS
- High lot-to-lot consistency
- Low endotoxin and hemoglobin levels
Identifiants chimiques
| Manufactured in cGMP (21 CFR 820) compliant and ISO9001 certified facilities | |
| United States | |
| Sterile | |
| ≤ 10 EU/mL | |
| Typically less than 2 μg/mL | |
| No |
| Sera should be stored at -10°C or lower. Once thawed, sera should be stored at 2 to 8°C. | |
| FBS Replacement Serum | |
| Sequential-filtered, 100 nm | |
| ≤ 20 mg/dL | |
| HyClone | |
| USDA Inspected |
Spécifications
| Albumin, Alkaline Phosphatase, Blood Urea Nitrogen, Calcium, Chloride, Creatinine, Gamma Globulin, Glucose, IgG, Inorganic Phosphorus, Iron, Lactate Dehydrogenase, Osmolality, Percent Iron Saturation, pH, Potassium, SGOT, SGPT, Sodium, TIBC, Total Bilirubin, Total Protein | |
| Sera should be stored at -10°C or lower. Once thawed, sera should be stored at 2 to 8°C. | |
| FBS Replacement Serum | |
| Sterile | |
| None | |
| ≤ 10 EU/mL | |
| ≤ 20 mg/dL | |
| HyClone | |
| USDA Inspected |
| Manufactured in cGMP (21 CFR 820) compliant and ISO9001 certified facilities | |
| United States | |
| 500 mL | |
| Sequential-filtered, 100 nm | |
| Bluetongue, Bovine Adenovirus, Bovine Parvovirus, Bovine RSV, BVDV, Cytopathogenic Agents, Hemadsorbing Agents, Rabies Virus, Reovirus | |
| No | |
| Typically less than 2 μg/mL | |
| No |