missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HyClone™ FetalClone™ I Serum (U.S.)

Numéro de catalogue. p-7058048
Change view
Click to view available options
Quantity:
100 mL
500 mL
Heat Inactivated:
No
Yes
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity Heat Inactivated
SH3008002 100 mL No
SH3008003 500 mL No
SH3008002HI 100 mL Yes
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Numéro de catalogue. SH3008002 Fournisseur Cytiva Code fournisseur SH30080.02
Il en reste null

Optimized for growth of hybridomas, supplement cell culture growth with U.S.-sourced HyClone™ FetalClone™ I Serum, a fetal bovine serum (FBS) alternative containing comparable IgG levels to those found in FBS.

  • Economical alternative to FBS
  • Performance comparable with FBS
  • High lot-to-lot consistency
  • Low endotoxin and hemoglobin levels
  • Demonstrated performance with a variety of cell lines, including hybridomas, CHO, BHK-21, NS0, MRC-5, and Vero cells
  • Spécifications

    Additional Assays Albumin, Alkaline Phosphatase, Blood Urea Nitrogen, Calcium, Chloride, Creatinine, Gamma Globulin, Glucose, IgG, Inorganic Phosphorus, Iron, Lactate Dehydrogenase, Osmolality, Percent Iron Saturation, pH, Potassium, SGOT, SGPT, Sodium, TIBC, Total Bilirubin, Total Protein
    Certifications/Compliance Manufactured in cGMP (21 CFR 820) compliant and ISO9001 certified facilities
    Content And Storage Sera should be stored at -10°C or lower. Once thawed, sera should be stored at 2° to 8°C.
    Description FetalClone I Serum (U.S.)
    Country of Origin United States
    Product Type FBS Replacement Serum
    Quantity 100 mL
    Sterility Sterile
    Sterilization Method Sequential-filtered, 100 nm
    Tested For Bluetongue, Bovine Adenovirus, Bovine Parvovirus, Bovine RSV, BVDV, Cytopathogenic Agents, Hemadsorbing Agents, Rabies Virus, Reovirus
    Endotoxin Concentration ≤ 10 EU/mL
    Heat Inactivated No
    Hemoglobin Concentration ≤ 20 mg/dL
    IgG levels Typically less than 2μg/mL
    Mycoplasma Detected No
    USDA Safety Testing Included USDA Inspected
    Afficher plus de résultats Afficher moins de résultats