missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HyClone™ Fetal Bovine Serum, Super Low IgG

Catalog No. SH983IH2540
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SH983IH2540 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SH983IH2540 Supplier Cytiva Supplier No. SH30898.03IH2540
NLSU: Get 1 Consumable Product When You Buy 1 Obtenez 1 produit consommable à l’achat d’un produit
View all promotions

Enhances cell culture growth, processed via protein G chromatography to substantially reduce immunoglobulin G (IgG) levels

Specifications

Country of Origin United States
Form Serum
Product Type Fetal Bovine Serum
Quantity 500 mL
Grade Super Low IgG
Processing Heat Inactivated, Irradiated (25 to 40 kGy)
Volume (Metric) 500 mL
For Use With (Application) Enhanceing Cell Culture Growth
Heat Inactivated Yes
IgG levels Super Low
Product Line HyClone
Show More Show Less