missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HyClone™ Fetal Bovine Serum, Super Low IgG

Enhances cell culture growth, processed via protein G chromatography to substantially reduce immunoglobulin G (IgG) levels

$276.38 - $1,666.69

Specifications

Country of Origin United States
Product Type Fetal Bovine Serum
Sterility Sterile
Endotoxin Concentration ≤ 10 EU/mL
For Use With (Application) Monoclonal Antibody Production, Virus Propagation, and Immunoassay Procedures
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Processing Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Processing Price Quantity & Availability  
SH3089801
View Documents Promotion Details
Cytiva
SH30898.01
50 mL None
Each for $276.38
Only null left
 
SH3089802HI
View Documents Promotion Details
Cytiva
SH30898.02HI
100 mL Heat Inactivated
Each for $409.27
Only null left
 
SH3089803HI
View Documents Promotion Details
Cytiva
SH30898.03HI
500 mL Heat Inactivated
Each for $1,666.69
Only null left
 
Description

Description

  • Processed using protein G chromatography to reduce levels to those required for monoclonal antibody production
  • Designed for applications where extremely low levels of inherent bovine IgG are required
  • Chromatographically treated to reduce IgG levels to less than 5 μg/mL, while still retaining excellent cell growth properties
  • For use in monoclonal antibody production, virus propagation, and immunoassay procedures
Specifications

Specifications

United States
Sterile
Monoclonal Antibody Production, Virus Propagation, and Immunoassay Procedures
≤ 5 μg/mL
Fetal Bovine Serum
≤ 10 EU/mL
≤ 10 mg/dL
HyClone
Videos
SDS
Documents

Documents