missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HyClone™ Fetal Bovine Serum, Super Low IgG

Catalog No. SH3089801
Change view
Click to view available options
Quantity:
50 mL
100 mL
500 mL
Processing:
Heat Inactivated
None
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Processing
SH3089801 50 mL None
SH3089802HI 100 mL Heat Inactivated
SH3089803HI 500 mL Heat Inactivated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. SH3089801 Supplier Cytiva Supplier No. SH30898.01
Get 1 Consumable Product When You Buy 1 Obtenez 1 produit consommable à l’achat d’un produit
View all promotions

Enhances cell culture growth, processed via protein G chromatography to substantially reduce immunoglobulin G (IgG) levels

  • Processed using protein G chromatography to reduce levels to those required for monoclonal antibody production
  • Designed for applications where extremely low levels of inherent bovine IgG are required
  • Chromatographically treated to reduce IgG levels to less than 5 μg/mL, while still retaining excellent cell growth properties
  • For use in monoclonal antibody production, virus propagation, and immunoassay procedures

Specifications

Content And Storage < -10°C
Country of Origin United States
Form Liquid
Product Type Fetal Bovine Serum
Quantity 50 mL
Sterility Sterile
Sterilization Method Sequential-filtered, 40 nm
Processing None
Endotoxin Concentration ≤ 10 EU/mL
For Use With (Application) Monoclonal Antibody Production, Virus Propagation, and Immunoassay Procedures
Heat Inactivated No
Hemoglobin Concentration ≤ 10 mg/dL
IgG levels ≤ 5 μg/mL
Product Line HyClone
Show More Show Less