missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HyClone™ Characterized Fetal Bovine Serum (New Zealand)

Catalog No. SH3462IR253
Change view
Click to view available options
Quantity:
500 mL
Processing:
Irradiated (25 to 35 doses)
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Processing
SH3462IR253 500 mL Irradiated (25 to 35 doses)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SH3462IR253 Supplier Cytiva Supplier No. SH30406.02IR2535
Get 1 Consumable Product When You Buy 1 Obtenez 1 produit consommable à l’achat d’un produit
View all promotions

Enhance growth of cells with New Zealand sourced HyClone™ Characterized Fetal Bovine Serum.

  • HyClone Characterized Fetal Bovine Serum (FBS), New Zealand Origin is carefully collected, processed, and filtered in our own New Zealand facility.
  • The great expanse of the Pacific ocean has protected New Zealand’s islands from many outside influences, both geographically and biologically.
  • One of the benefits of this isolation is that New Zealand has the fewest reported bovine diseases in the world, making Characterized FBS, New Zealand Origin an excellent source of bovine serum as it offers greater security.
  • Characterized FBS, New Zealand Origin is filtered through three sequential 100 nm (0.1 μm) pore size-rated filters.
  • Before dispensing, each lot of serum is pooled to ensure uniformity and consistency between bottles (true pool technology).

Specifications

Additional Assays Progesterone, Chloride, Insulin, Inorganic Phosphorus, PTH, Glucose, Cholesterol, Copper, HDL, Magnesium, LDL, Selenium, Triglycerides, IgG, Gamma Globulin, pH, Phospholipids, Osmolality, Alkaline Phosphatase, Iron, Lactate Dehydrogenase, TIBC, SGPT, Percent Saturation, SGOT, B1, B2, B6, Niacin, Biotin, Retinol, Carotene, Tocopherol, B12, Folate, Vitamin C, 1, 25 Dihydroxy Vitamin D, Total Protein, Albumin, and Blood Urea Nitrogen
Content And Storage Store at or below -10°C
Country of Origin New Zealand
Form Liquid
Product Type Fetal Bovine Serum
Quantity 500 mL
Sterility Sterile
Sterilization Method Sequential-filtered, 100 nm
Grade Characterized
Processing Irradiated (25 to 35 doses)
Tested For Bluetongue, Bovine Adenovirus, Bovine Parvovirus, Bovine RSV, BVDV, Cytopathogenic Agents, Hemadsorbing Agents, Rabies Virus and Reovirus
Endotoxin Concentration ≤ 20 EU/mL
Heat Inactivated No
Hemoglobin Concentration ≤ 25 mg/dL
Product Line HyClone
USDA Safety Testing Included USDA Inspected
Show More Show Less