missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HyClone™ Characterized Fetal Bovine Serum (FBS), U.S. Origin

Catalog No. SH103IH2540
Change view
Click to view available options
Quantity:
500 mL
Processing:
HI, Irr - 25 to 40 doses
HI, Irr - 30 to 45 doses
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Processing
SH103IH2540 500 mL HI, Irr - 25 to 40 doses
SH103IH3045 500 mL HI, Irr - 30 to 45 doses
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. SH103IH2540 Supplier Cytiva Supplier No. SH30071.03IH2540

Low in antibodies and high in growth factors to support robust growth and division of cells, including embryonic stem cells.

  • Additional traceability testing of HyClone™ Fetal Bovine Serum is performed by a collaborator, Oritain
  • Low in antibodies and high in growth factors
  • Filtered through three sequential 100 nm (0.1 μm) pore size-rated filters
  • Each lot is subjected to virus panel testing according to 9 CFR 113.53
  • Characterized FBS is assayed for many components, including gamma globulin, glucose, and IgG
  • Supports robust growth and division of cells, including embryonic stem cells
  • Produced using true pool technology to minimize bottle-to-bottle variability
  • Complete traceability is provided back to the original source, as certified by the International Serum Industry Association (ISIA)
  • Sourced in the USA

Specifications

Country of Origin United States
Form Serum
Product Type Fetal Bovine Serum
Quantity 500 mL
Sterility Sterile
Sterilization Method Sequential Triple-filtered, 100 nm
Grade Characterized
Processing HI, Irr - 25 to 40 doses
Tested For Virus Panel Testing According to 9 CFR 113.53
Volume (Metric) 500 mL
Product Line HyClone
Show More Show Less