missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HyClone™ Calf Serum (New Zealand)

Catalog No. SH3040102
Change view
Click to view available options
Quantity:
1000 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SH3040102 1000 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SH3040102 Supplier Cytiva Supplier No. SH30401.02

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Supplement cell culture growth with New Zealand-sourced HyClone™ Bovine Calf Serum, an excellent and cost-effective alternative to fetal bovine serum (FBS).

Specifications

Additional Assays Albumin, Alkaline Phosphatase, Blood Urea Nitrogen, Calcium, Chloride, Creatinine, Gamma Globulin, Glucose, IgG, Inorganic Phosphorus, Iron, Lactate Dehydrogenase, Osmolality, Percent Saturation, pH, Potassium, SGOT, SGPT, Sodium, TIBC, Total Bilirubin, Total Protein
Certifications/Compliance USDA Regulations
Content And Storage Sera should be stored at -10°C or lower. Once thawed, sera should be stored at 2° to 8°C.
Description Calf Serum (New Zealand)
Quantity 1000 mL
Sterility Sterile
Sterilization Method 0.1 μm filtration
Tested For Bluetongue, Bovine Adenovirus, Bovine Parvovirus, Bovine RSV, BVDV, Cytopathogenic Agents, Hemadsorbing Agents, Rabies Virus, Reovirus
Endotoxin Concentration ≤ 25 EU/mL
Mycoplasma Detected Yes
USDA Safety Testing Included Yes
Show More Show Less