missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ HybridSPE™-Phospholipid Cartridge

Catalog No. 111009512
Change view
Click to view available options
Bed Weight:
30 mg
500 mg
Volume (Metric):
1 mL
6 mL
Quantity:
100/Pk.
200/Pk.
30/Pk.
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Bed Weight Volume (Metric) Quantity
111009512 30 mg 1 mL 100/Pk.
111008743 30 mg 1 mL 200/Pk.
111009120 500 mg 6 mL 30/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 111009512 Supplier MilliporeSigma™ Supplier No. 55261U

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

HybridSPE™ Phospholipid Removal Technology for Biological Matrices. Remove phospholipids and proteins for accurate and reproducible LC-MS analysis.

Key Features and Benefits:

  • More accurate results
  • Unique chemistry can effectively separate hydrophobic analytes (such as vitamins) from phospholipids, unlike competitive product chemistries
  • Elimination of proteins and phospholipid-induced ion suppression
  • Simultaneous removal of proteins and phospholipids
  • Simple, standardized methodology, analogous to traditional protein precipitation
  • Alternative to complex traditional SPE method development
  • Reproducible, consistent performance reduces need for reprocessing
  • Less instrument downtime and longer column life
  • Decreased run times by eliminating the need for gradients to clean columns between samples
  • High throughput processing that is automatable and compatible with common robotic systems
  • Ready-to-use, no preconditioning required

Specifications

Bed Weight 30 mg
For Use With (Application) Food and Beverage
Type Fritless SPE Cartridge and Plates
Volume (Metric) 1 mL
Quantity 100/Pk.