missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ HYAL3 Polyclonal Antibody
GREENER_CHOICE

Numéro de catalogue. PIPA579422
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
PIPA579422 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. PIPA579422 Fournisseur Invitrogen™ Code fournisseur PA579422
Il en reste null

Rabbit Polyclonal Antibody

Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: 22RV Cell, HELA Cell, V20S Cell. IHC: Human Mammary Cancer Tissue.

This gene encodes a member of the hyaluronidase family. Hyaluronidases are endoglycosidase enzymes that degrade hyaluronan, one of the major glycosaminoglycans of the extracellular matrix. The regulated turnover of hyaluronan plays a critical role in many biological processes including cell proliferation, migration and differentiation. The encoded protein may also play an important role in sperm function. This gene is one of several related genes in a region of chromosome 3p21.3 associated with tumor suppression, and the expression of specific transcript variants may be indicative of tumor status. Alternatively spliced transcript variants encoding multiple isoforms have been observed for this gene, and some isoforms may lack hyaluronidase activity. This gene overlaps and is on the same strand as N-acetyltransferase 6 (GCN5-related), and some transcripts of each gene share a portion of the first exon.

Spécifications

Antigen HYAL3
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 500 μg/mL
Conjugate Unconjugated
Formulation PBS with 5mg BSA and 0.05mg sodium azide, 0.05mg thimerosal
Gene HYAL3
Gene Accession No. O43820, Q8VEI3
Gene Alias Hyal3; hyal-3; Hyaluronidase; hyaluronidase 3; hyaluronidase-3; hyaluronoglucosaminidase 3; hyaluronoglucosaminidase-3; Hyl3; LUCA14; LUCA3; LuCa-3; Lung carcinoma protein 3; Minna14
Gene Symbols HYAL3
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence at the C-terminus of human HYAL3 (357-371aa SHQRCHGHGRCARRD).
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 109685, 8372
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Isotype IgG
Afficher plus de résultats Afficher moins de résultats