missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ HYAL1 Polyclonal Antibody
GREENER_CHOICE

Numéro de catalogue. PIPA579420
Modifier l'affichage
Click to view available options
Quantité:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantité
PIPA579420 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. PIPA579420 Fournisseur Invitrogen™ Code fournisseur PA579420
Saison des économies
Afficher toutes les promotions

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Rabbit Polyclonal Antibody

Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: human HepG2 whole cell, human PC-3 whole cell, monkey COS-7 whole cell, rat liver tissue, mouse liver tissue, mouse kidney tissue. IHC: Human Intestinal Cancer tissue IHC-F: Human Placenta tissue. ICC/IF: MCF-7 cell.

This gene encodes a lysosomal hyaluronidase. Hyaluronidases intracellularly degrade hyaluronan, one of the major glycosaminoglycans of the extracellular matrix. Hyaluronan is thought to be involved in cell proliferation, migration and differentiation. This enzyme is active at an acidic pH and is the major hyaluronidase in plasma. Mutations in this gene are associated with mucopolysaccharidosis type IX, or hyaluronidase deficiency. The gene is one of several related genes in a region of chromosome 3p21.3 associated with tumor suppression. Multiple transcript variants encoding different isoforms have been found for this gene.

Spécifications

Antigène HYAL1
Applications Immunocytochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 500 μg/mL
Conjugué Unconjugated
Formule PBS with 5mg BSA and 0.05mg thimerosal, 0.05mg sodium azide
Gène HYAL1
Numéro d’acquisition du gène Q12794, Q76HN1, Q91ZJ9
Alias du gène Hya1; HYAL1; Hyal-1; Hyaluronidase; hyaluronidase 1; hyaluronidase-1; hyaluronoglucosaminidase 1; hyaluronoglucosaminidase-1; LUCA1; LuCa-1; Lung carcinoma protein 1; MPS9; NAT6; plasma hyaluronidase; tumor suppressor LUCA-1
Symboles de gène HYAL1
Espèces hôtes Rabbit
Immunogène A synthetic peptide corresponding to a sequence at the N-terminus of human HYAL1 (32-47aa FTTVWNANTQWCLERH).
Méthode de purification Antigen affinity chromatography
Quantité 100 μg
Statut réglementaire RUO
Primaire ou secondaire Primary
ID du gène (Entrez) 15586, 3373, 367166
Espèces cibles Human, Mouse, Monkey, Rat
Contenu et stockage -20°C
Type de produit Antibody
Forme Lyophilized
Isotype IgG
Afficher plus de résultats Afficher moins de résultats