Learn More
Fisherbrand™ Humidity and Temperature Data Logger
Ultra-high speed download capability
Spécifications
| Type | Temperature and Humidity Datalogger |
|---|---|
| Data Memory | 1,000,000/Channel |
| Calibration | Digital Calibration |
| Alarm Types | High/Low |
| Battery Type | 3.6 V Lithium |
| Numéro de catalogue | Numéro du manufacturier. | Certifications/Compliance | Prix | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Numéro de catalogue | Numéro du manufacturier. | Certifications/Compliance | Prix | Quantity & Availability | |||||
|
15059306
|
Fisherbrand™
RHTEMP101A |
CE |
N/A
|
|
Connectez-vous ou enregistrez-vous pour vérifier votre prix et la disponibilité.
|
||||
Description
Ultra-high speed download capability
Using the data logging software, starting, stopping and downloading from the unit is simple and easy. Graphical, tabular and summary data is provided for analysis and data can be viewed in °C, °F, K,°R, %RH, mg/mL water vapor concentration and dew point. The data can also be automatically exported to Excel™ for further calculations. Free firmware upgrades for the life of the product are available, so that data loggers already deployed in the field can grow with the technology. Units do not need to be returned to the factory for upgrades. The user can do this automatically from any PC.
- 10 year battery life
- One-second reading rate
- Multiple start/stop function
- Ultra-high speed download capability
- 500,000 reading per channel storage capacity
- Optional memory wrap
- Battery life indicator
- Optional password protection
- Programmable high-low alarms
- Temperature sensor: precision RTD element
- Temperature range: -40°C to +80°C (-40°F to +176°F)
- Temperature resolution: 0.01°C (0.018°F)
- Temperature calibrated accuracy: ±0.5°C (±0.9°F)
- Humidity sensor: internal semiconductor
- Humidity range: 0ã95% RH
- Humidity resolution: 0.1% RH
- Humidity calibrated accuracy: ±3.0%RH (±2%RH typical at 25°C/77°F)
- RH Units: %RH, dew point, water vapor, concentration (mg/hl)
- Memory: 500,000 readings per channel; software configurable memory wrap; 250,000 readings in multiple start/stop mode
- Reading rate: 1 reading every second to 1 reading every 24 hours
- Alarm: Programmable high and low limits; activated when humidity reaches or exceeds set limits
- Baud rate: 115,200
- LED indicators: red and green
- Push-button start and stop: Yes
- Battery life: 10 years typical at a 15 minute reading rate (ambienbt temp.)
- Material: ABS plastic
- Dimensions: 1.4 x 2.2 x 0.6 in. (3.6 x 5.6 x 16cm)
- Operating environment: -40°Cã+80°C (-40°F to +176°F), 0ã95%RH non-condensing
- Required interface package: IFC200
- Weight: 0.0563 lb.
Order Info
Software, Manual and USB Interface Cable Package required (p/n 15-060-036).
Compliance
CE approved.
Spécifications
| Temperature and Humidity Datalogger | |
| Digital Calibration | |
| 3.6 V Lithium | |
| IFC200, USB | |
| RHTEMP101A | |
| Precision RTD Element (Temperature), Internal Semiconductor (Humidity) | |
| 0 to 95% | |
| -40°C to +80°C | |
| -40°C to +80°C | |
| 24 g |
| 1,000,000/Channel | |
| High/Low | |
| 10 Years | |
| ABS Plastic | |
| Software, Manual, USB Interface Cable Package Required (p/n 15-060-036) | |
| For implementation of HACCP programs, HVAC, warehouse monitoring, museum monitoring, medical and pharmaceutical applications | |
| 0.01°C | |
| -40°F to +176°F | |
| 0.9 oz. |