missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human USP27X (aa 416-438) Control Fragment Recombinant Protein

Numéro de catalogue. RP108110
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
RP108110 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. RP108110 Fournisseur Invitrogen™ Code fournisseur RP108110
Il en reste null

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (83%), Rat (83%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-67449 (PA5-67449. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

Deubiquitinase that can reduce the levels of BCL2L11/BIM ubiquitination and stabilize BCL2L11 in response to the RAF-MAPK-degradation signal. By acting on BCL2L11 levels, may counteract the anti-apoptotic effects of MAPK activity.

Spécifications

Accession Number A6NNY8
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 389856
Name Human USP27X (aa 416-438) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias Deubiquitinating enzyme 27; RGD1564628; Sfc11; Ubiquitin carboxyl-terminal hydrolase 22-like; Ubiquitin carboxyl-terminal hydrolase 27; ubiquitin carboxyl-terminal hydrolase 51-like; ubiquitin specific peptidase 27 X-linked; ubiquitin specific peptidase 27, x chromosome; ubiquitin specific peptidase 27, X-linked; ubiquitin specific peptidase 27, X-linked, pseudogene 1; ubiquitin specific protease 27, x chromosome; ubiquitin thioesterase 27; ubiquitin thiolesterase 22; ubiquitin thiolesterase 27; ubiquitin-specific-processing protease 27; USP22L; USP27; Usp27x; Usp27x-ps1; X-linked ubiquitin carboxyl-terminal hydrolase 27; X-linked ubiquitin thioesterase 27
Common Name USP27X
Gene Symbol USP27X
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence LFYHKQVLEHESEKVKEMNTQAY
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Afficher plus de résultats Afficher moins de résultats