missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human TNF-alpha (Research Grade Adalimumab Biosimilar) Alexa Fluor 594-conjugated Antibody, R&D Systems™
Description
TNF-alpha Monoclonal specifically detects TNF-alpha in Human samples. It is validated for Flow Cytometry (Intracellular Staining).
Spécifications
Spécifications
| Antigène | TNF-alpha |
| Applications | Flow Cytometry (Intracellular Staining) |
| Classification | Monoclonal |
| Clone | Hu7 |
| Conjugué | Alexa Fluor 594 |
| Dilution | Intracellular Staining by Flow Cytometry 0.25-1μg/106 cells |
| Formule | Supplied 0.2Mg/mL in a saline solution containing BSA and Sodium Azide. |
| Alias du gène | APC1 protein, Cachectin, Cachetin, DIF, TNF, TNF, monocyte-derived, TNF-a, TNFA, TNFalpha, TNF-alphacachectin, TNFATNF, macrophage-derived, TNFG1F, TNFSF1A, TNFSF2, TNFSF2TNF superfamily, member 2, tumor necrosis factor, tumor necrosis factor (TNF superfamily, member 2), tumor necrosis factor alpha, Tumor necrosis factor ligand superfamily member 2, tumor necrosis factor-alpha |
| Espèces hôtes | Human |
| Immunogène | Human TNF-alpha |
| Afficher plus de résultats |