Learn More
Danaher Corp Human T Cell Exhaustion Kit
Numéro de catalogue. 502891678
Encompass
Speed: Sample to actionable data by allowing multiple assays to be run in one assay thereby reducing the hands-on time numbers of overall tests and conserving precious samples.Results: 11 endpoints from a single-well assay measuring T cell exhaustion phenotypes and function as well as cytokine secretion in response to stimulation cell viability and an optional proliferation dye allows screening for molecules that reverse T cell exhaustion.Biological insight: Generate functional data and analyze cell health from the same well by multiplexing cell and cytokine detection in one assay.Simplicity: Single platform assay data acquisition and analysis eliminate the time of acquiring and correlating data from multiple sources.
Numéro de catalogue. 502891678 Fournisseur Danaher Corp Code fournisseur 97071
Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.
Speed: Sample to actionable data by allowing multiple assays to be run in one assay thereby reducing the hands-on time numbers of overall tests and conserving precious samples.Results: 11 endpoints from a single-well assay measuring T cell exhaustion phenotypes and function as well as cytokine secretion in response to stimulation cell viability and an optional proliferation dye allows screening for molecules that reverse T cell exhaustion.Biological insight: Generate functional data and analyze cell health from the same well by multiplexing cell and cytokine detection in one assay.Simplicity: Single platform assay data acquisition and analysis eliminate the time of acquiring and correlating data from multiple sources.