missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human SPARC Alexa Fluor 647-conjugated Antibody, R&D Systems™
Description
SPARC Monoclonal specifically detects SPARC in Human samples. It is validated for Flow Cytometry (Intracellular Staining).
Spécifications
Spécifications
| Antigène | SPARC |
| Applications | Flow Cytometry (Intracellular Staining) |
| Classification | Monoclonal |
| Clone | 122511 |
| Concentration | 0.2 mg/mL |
| Conjugué | Alexa Fluor 647 |
| Dilution | Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells |
| Formule | Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide |
| Numéro d’acquisition du gène | P09486 |
| Alias du gène | Basement-membrane protein 40, BM-40, ONcysteine-rich protein, osteonectin, Secreted protein acidic and rich in cysteine, secreted protein, acidic, cysteine-rich (osteonectin) |
| Afficher plus de résultats |
For Research Use Only (RUO)